This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SECISBP2 SIRNA Assays ›
Gene Symbol:
Secisbp2
Gene Name:
SECIS binding protein 2
Gene Aliases:
2210413N07Rik, 2810012K13Rik, SBP2
Chromosome Location:
Chr.13: 51650829 - 51684044 on Build GRCm38
UniGene ID:
Mm.275981
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 508576

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Secisbp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK008918.1 - 5 820
AK030021.1 - 14 2110
AK156076.1 - 14 2186
AK168634.1 - 14 2143
BC038877.1 - 2 256
BC125391.1 - 14 2166 40129906
BC125393.1 - 14 2163 40129908
BC144795.1 - 14 2161

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline