This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KRT24 SIRNA Assays ›
Gene Symbol:
Krt24
Gene Name:
keratin 24
Gene Aliases:
2310058N18Rik, 2310075C18Rik
Chromosome Location:
Chr.11: 99280093 - 99285238 on Build GRCm38
UniGene ID:
Mm.46378
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 508695

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Krt24
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029393.1 NP_083669.1 1 536
GenBank mRNA AK010165.1 - 1 561
AK053678.1 - 1 282
BC129847.1 - 1 536 40107361
BC129848.1 - 1 536 40107363
GU046702.1 - 1 505

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
structural molecule activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0