siRNA Details
| Gene Symbol: Ppp1r18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175242.1 | NP_780451.1 | 1 | 462 | ||
| GenBank mRNA | AK051656.1 | - | 1 | 418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r18| Gene Symbol: Ppp1r18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175242.1 | NP_780451.1 | 1 | 462 | ||
| GenBank mRNA | AK051656.1 | - | 1 | 418 | ||