This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GRIFIN SIRNA Assays ›
Gene Symbol:
Grifin
Gene Name:
galectin-related inter-fiber protein
Gene Aliases:
E130101O17Rik
Chromosome Location:
Chr.5: 140563117 - 140567628 on Build GRCm38
UniGene ID:
Mm.160040
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 509096

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Grifin
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030022.1 NP_084298.1 2, 3 104
GenBank mRNA AK021365.1 - 2, 3 105
BC107323.2 - 2, 3 104 40053455
BC107324.1 - 2, 3 104 40053456

Back To Top

More Information


Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-qtvkm:80/100.66.79.40:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0