This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PRG4 SIRNA Assays ›
Gene Symbol:
Prg4
Gene Name:
proteoglycan 4 (megakaryocyte stimulating factor, articular superficial zone protein)
Gene Aliases:
CACP, DOL54, JCAP, MSF, SZP, lubricin
Chromosome Location:
Chr.1: 150449412 - 150466221 on Build GRCm38
UniGene ID:
Mm.174256
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 509247

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prg4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110146.1 NP_001103616.1 6 497
NM_021400.3 NP_067375.3 7 2891
GenBank mRNA AB034730.1 - 8 2390
AK132597.1 - 1 637
AK149469.1 - 6 498
BC130021.1 - 5 461

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune response
regulation of cell proliferation
negative regulation of interleukin-6 biosynthetic process
hematopoietic stem cell proliferation
scavenger receptor activity
polysaccharide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-x6g4t:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline