This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SYT5 SIRNA Assays ›
Gene Symbol:
Syt5
Gene Name:
synaptotagmin V
Gene Aliases:
SytIX
Chromosome Location:
Chr.1: 72860218 - 72867934 on Build Rnor_6.0
UniGene ID:
Rn.11030
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 51021

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Syt5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019350.2 NP_062223.1 4 627
XM_006228287.2 XP_006228349.1 3 317
GenBank mRNA BC092198.1 - 4 627
U26402.1 - 4 430
X84884.1 - 3 380

Back To Top

More Information


Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

calcium ion regulated exocytosis
transporter activity
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline