This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PRL8A3 SIRNA Assays ›
Gene Symbol:
Prl8a3
Gene Name:
prolactin family 8, subfamily a, member 3
Gene Aliases:
Plp-Cv, Plpc, Plpcv, Prlpc, Prlpc1
Chromosome Location:
Chr.17: 39455865 - 39463715 on Build Rnor_6.0
UniGene ID:
Rn.202972
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 51068

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prl8a3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020079.1 NP_064464.1 317
GenBank mRNA U93351.1 - 317

Back To Top

More Information


Panther Classification:

Molecular Function -

peptide hormone

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gmb9z:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline