This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IP6K2 SIRNA Assays ›
Gene Symbol:
Ip6k2
Gene Name:
inositol hexakisphosphate kinase 2
Gene Aliases:
Ihpk2, Pius
Chromosome Location:
Chr.8: 117574104 - 117599849 on Build Rnor_6.0
UniGene ID:
Rn.112574
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 51216

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ip6k2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021660.2 NP_067692.2 3 404
XM_008766564.1 XP_008764786.1 2 370
XM_008766566.1 XP_008764788.1 2 370
GenBank mRNA BC083574.1 - 3 404 7112777

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029634, AK057615, AK005166, BC039922, AK050593, and others.

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphate ion transport
negative regulation of cell growth
positive regulation of apoptotic process
phosphatidylinositol phosphorylation
inositol hexakisphosphate 5-kinase activity
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-clzgt:80/100.66.76.159:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline