siRNA Details
| Gene Symbol: Nop58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021754.1 | NP_068522.1 | 2 | 249 | ||
| GenBank mRNA | AF194371.1 | - | 2 | 249 | ||
| BC087637.1 | - | 2 | 282 | 7309062 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nop58| Gene Symbol: Nop58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021754.1 | NP_068522.1 | 2 | 249 | ||
| GenBank mRNA | AF194371.1 | - | 2 | 249 | ||
| BC087637.1 | - | 2 | 282 | 7309062 | ||