This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PLRG1 SIRNA Assays ›
Gene Symbol:
Plrg1
Gene Name:
pleiotropic regulator 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 182040338 - 182056669 on Build Rnor_6.0
UniGene ID:
Rn.44181
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 51268

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Plrg1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021757.2 NP_068525.1 3 299
GenBank mRNA AF128241.1 - 2 206
BC087742.1 - 3 299 7314208
FQ215141.1 - 3 319

Back To Top

More Information


Gene Ontology Categories:

Function(s)

mRNA processing
RNA splicing
protein localization to nucleus
positive regulation of G1/S transition of mitotic cell cycle

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline