siRNA Details
| Gene Symbol: Insrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022212.2 | NP_071548.2 | 2 | 841 | ||
| XM_006232749.2 | XP_006232811.2 | 2 | 911 | |||
| XM_008761207.1 | XP_008759429.1 | 2 | 911 | |||
| GenBank mRNA | D13965.1 | - | 2 | 443 | ||
| D13966.1 | - | 2 | 443 | |||
| M90661.1 | - | 2 | 841 | |||

Insrr
Genomic Map