siRNA Details
| Gene Symbol: DMPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081560.2 | NP_001075029.1 | 2 | 416 | ||
| NM_001081562.2 | NP_001075031.1 | 2 | 416 | |||
| NM_001081563.2 | NP_001075032.1 | 1 | 785 | |||
| NM_001288764.1 | NP_001275693.1 | 3 | 475 | |||
| NM_001288766.1 | NP_001275695.1 | 2 | 416 | |||
| NM_004409.4 | NP_004400.4 | 2 | 416 | |||
| GenBank mRNA | AB209363.1 | - | 2 | 1259 | ||
| AK302874.1 | - | 2 | 349 | |||
| AK304749.1 | - | 1 | 688 | |||
| BC026328.1 | - | 3 | 470 | 4792104 | ||
| BC062553.1 | - | 2 | 337 | 6480902 | ||
| HY031977.1 | - | 2 | 359 | |||
| KJ534826.1 | - | 1 | 241 | |||
| KJ534827.1 | - | 1 | 241 | |||
| KJ534828.1 | - | 1 | 241 | |||
| L19268.1 | - | 2 | 338 | |||
| M87312.1 | - | 1 | 786 | |||
| M94203.1 | - | 2 | 88 | |||
| S72883.1 | - | 2 | 247 | |||

DMPK
Genomic Map