siRNA Details
| Gene Symbol: Vax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022636.1 | NP_072158.1 | 3 | 479 | ||
| GenBank mRNA | AF113515.1 | - | 3 | 479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vax1| Gene Symbol: Vax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022636.1 | NP_072158.1 | 3 | 479 | ||
| GenBank mRNA | AF113515.1 | - | 3 | 479 | ||