This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EPHA2 SIRNA Assays ›
Gene Symbol:
EPHA2
Gene Name:
EPH receptor A2
Gene Aliases:
ARCC2, CTPA, CTPP1, CTRCT6, ECK
Chromosome Location:
Chr.1: 16124337 - 16156104 on Build GRCh38
UniGene ID:
Hs.171596
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 521

Purification
Size
Availability Inventoried
Catalog # AM51331
Price 6,301.00
Your Price
Online offer:
6301.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: EPHA2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017000534.1 XP_016856023.1 2, 3 429
XM_017000535.1 XP_016856024.1 2, 3 389
XM_017000537.1 XP_016856026.1 2, 3 331
GenBank mRNA BC037166.2 - 2, 3 277 5090654
CB991812.1 - 2, 3 341
EU826606.1 - 2, 3 159
HY005615.1 - 2, 3 337
M59371.1 - 2, 3 272

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010139, U07634, CJ088701, AK144202, BC140960, and others.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
angiogenesis
vasculogenesis
osteoblast differentiation
multicellular organism development
intrinsic apoptotic signaling pathway in response to DNA damage
regulation of lamellipodium assembly
notochord formation
viral process
cell migration
peptidyl-tyrosine phosphorylation
neural tube development
neuron differentiation
keratinocyte differentiation
osteoclast differentiation
mammary gland epithelial cell proliferation
regulation of cell adhesion mediated by integrin
post-anal tail morphogenesis
protein kinase B signaling
regulation of blood vessel endothelial cell migration
regulation of angiogenesis
bone remodeling
ephrin receptor signaling pathway
axial mesoderm formation
negative regulation of protein kinase B signaling
notochord cell development
cell chemotaxis
branching involved in mammary gland duct morphogenesis
lens fiber cell morphogenesis
regulation of ERK1 and ERK2 cascade
response to growth factor
positive regulation of establishment of protein localization to plasma membrane
activation of GTPase activity
cell-cell adhesion
transmembrane receptor protein tyrosine kinase activity
ephrin receptor activity
protein binding
ATP binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0