This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC100911854 SIRNA Assays ›
Gene Symbol:
LOC100911854 Mbl2
Gene Name:
mannose-binding protein C-like
mannose-binding lectin (protein C) 2
Gene Aliases:
-
Ab2-001, Ab2-011
Chromosome Location:
Chr.1: 248723370 - 248730969 on Build Rnor_6.0
Chromosome Location:
Chr.1: 248435069 - 248442669 on Build Rnor_6.0
UniGene ID:
-
Rn.9667
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 52435

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100911854
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749087.3 XP_003749135.1 779
XM_003749088.3 XP_003749136.1 699
Gene Symbol: Mbl2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022704.2 NP_073195.2 4 503
XM_006231249.2 XP_006231311.1 6 779
XM_006231250.2 XP_006231312.1 6 699
GenBank mRNA AY325174.1 - 533
AY325178.1 - 533
FQ210356.1 - 6 688
M14103.1 - 6 673
X05023.1 - 4 503

Back To Top

More Information


Panther Classification:

Molecular Function -

surfactant

Gene Ontology Categories:

Function(s) Process(es)

complement activation, lectin pathway
complement activation, classical pathway
positive regulation of protein processing
negative regulation of growth of symbiont in host
innate immune response
positive regulation of complement activation
negative regulation of viral process
positive regulation of phagocytosis
defense response to Gram-positive bacterium
protein oligomerization
protein homooligomerization
killing by host of symbiont cells
protein homotrimerization
protease binding
receptor binding
calcium ion binding
galactose binding
mannose binding
carbohydrate binding
protein self-association
calcium-dependent protein binding
phosphatidylinositol-4-phosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline