This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TAS2R107 SIRNA Assays ›
Gene Symbol:
Tas2r107
Gene Name:
taste receptor, type 2, member 107
Gene Aliases:
T2R4, Tas2r10
Chromosome Location:
Chr.4: 165773200 - 165774126 on Build Rnor_6.0
UniGene ID:
Rn.48784
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 52659

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tas2r107
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023995.1 NP_076485.1 1 625
GenBank mRNA AF227142.1 - 1 625

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
G-protein coupled receptor signaling pathway
biological_process
sensory perception of taste
detection of chemical stimulus involved in sensory perception of taste
G-protein coupled receptor activity
taste receptor activity
bitter taste receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0