siRNA Details
| Gene Symbol: Senp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023989.1 | NP_076479.1 | 6 | 581 | ||
| XM_008768805.1 | XP_008767027.1 | 6 | 712 | |||
| GenBank mRNA | AF260129.1 | - | 6 | 581 | ||
| FQ210639.1 | - | 6 | 676 | |||
| FQ212134.1 | - | 6 | 676 | |||
| FQ222500.1 | - | 5 | 524 | |||

Senp2
Genomic Map