This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GNB1 SIRNA Assays ›
Gene Symbol:
Gnb1
Gene Name:
guanine nucleotide binding protein (G protein), beta polypeptide 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 172913898 - 172981403 on Build Rnor_6.0
UniGene ID:
Rn.126047
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53151

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gnb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030987.2 NP_112249.2 6, 7 423
XM_006239517.2 XP_006239579.1 6, 7 575
XM_006239518.2 XP_006239580.1 6, 7 575
XM_006239519.2 XP_006239581.1 6, 7 575
XM_006239520.2 XP_006239582.1 6, 7 575
XM_008764335.1 XP_008762557.1 5, 6 458
GenBank mRNA AY552805.1 - 4, 5 275
BC078809.1 - 6, 7 423 7123318
DQ120483.1 - 1, 2 28
DQ120484.1 - 1, 2 28
U34958.1 - 4, 5 275
U88324.1 - 4, 5 312

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001160017, NM_008142, NM_001160016, AK198923, AK180913, and others.

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
metabolic process
cell proliferation
cardiac muscle cell apoptotic process
sensory perception of taste
retina development in camera-type eye
cellular response to hypoxia
G-protein coupled receptor binding
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline