siRNA Details
| Gene Symbol: Cd200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031518.2 | NP_113706.2 | 3 | 246 | ||
| XM_006248325.2 | XP_006248387.1 | 3 | 257 | |||
| GenBank mRNA | BC089793.1 | - | 3 | 246 | 7380792 | |
| X01785.1 | - | 3 | 203 | |||

Cd200
Genomic Map