siRNA Details
| Gene Symbol: Jak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031514.1 | NP_113702.1 | 4, 5 | 514 | ||
| GenBank mRNA | AJ000557.1 | - | 1, 2 | 31 | ||
| U13396.1 | - | 4, 5 | 514 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jak2| Gene Symbol: Jak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031514.1 | NP_113702.1 | 4, 5 | 514 | ||
| GenBank mRNA | AJ000557.1 | - | 1, 2 | 31 | ||
| U13396.1 | - | 4, 5 | 514 | |||