siRNA Details
| Gene Symbol: Nr1h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031627.2 | NP_113815.2 | 2 | 183 | ||
| XM_006234533.2 | XP_006234595.1 | 3 | 361 | |||
| XM_006234534.1 | XP_006234596.1 | 3 | 302 | |||
| XM_006234535.2 | XP_006234597.1 | 3 | 503 | |||
| XM_006234537.1 | XP_006234599.1 | 3 | 240 | |||
| XM_008762013.1 | XP_008760235.1 | 3 | 360 | |||
| GenBank mRNA | BC088750.1 | - | 2 | 183 | 7314121 | |
| U11685.1 | - | 2 | 190 | |||

Nr1h3
Genomic Map