siRNA Details
| Gene Symbol: MTMR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004687.4 | NP_004678.3 | 3, 4 | 196 | ||
| XM_005257784.2 | XP_005257841.1 | 2, 3 | 345 | |||
| XM_005257785.4 | XP_005257842.1 | 4, 5 | 313 | |||
| XM_005257786.4 | XP_005257843.1 | 3, 4 | 1100 | |||
| XM_006722168.3 | XP_006722231.1 | 3, 4 | 140 | |||
| XM_011525460.2 | XP_011523762.1 | 4, 5 | 1193 | |||
| GenBank mRNA | AB014547.2 | - | 1, 2 | 86 | ||
| AF264717.1 | - | 3, 4 | 139 | |||
| AK308877.1 | - | 2, 3 | 345 | |||
| BC035609.1 | - | 3, 4 | 157 | 5201989 | ||
| DR003190.1 | - | 3, 4 | 217 | |||

MTMR4
Genomic Map