This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC29A2 SIRNA Assays ›
Gene Symbol:
Slc29a2 LOC100911721
Gene Name:
solute carrier family 29 (equilibrative nucleoside transporter), member 2
equilibrative nucleoside transporter 2-like
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 220399911 - 220407935 on Build Rnor_6.0
Chromosome Location:
Chr.1: 220306607 - 220314631 on Build Rnor_6.0
UniGene ID:
Rn.37517
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 54140

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc29a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031738.1 NP_113926.1 8 997
XM_008760174.1 XP_008758396.1 2750
GenBank mRNA AF015305.1 - 8 997
Gene Symbol: LOC100911721
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749035.3 XP_003749083.1 8 1781
XM_008760194.1 XP_008758416.1 8 2750

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

lactation
adenine transport
guanine transport
nucleoside transport
uridine transport
cellular response to insulin stimulus
hypoxanthine transport
thymine transport
nucleoside transmembrane transport
nucleoside transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0