This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other USF1 SIRNA Assays ›
Gene Symbol:
Usf1
Gene Name:
upstream transcription factor 1
Gene Aliases:
-
Chromosome Location:
Chr.13: 89797750 - 89805561 on Build Rnor_6.0
UniGene ID:
Rn.10845
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 54263

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Usf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031777.2 NP_113965.1 4, 5 313
XM_006250266.2 XP_006250328.1 4, 5 498
GenBank mRNA AF026476.1 - 2, 3 182
BC088849.1 - 4, 5 313 7313699

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_007122, NM_001276373, BC035505, BP280296, AK314876, and others.
This siRNA also targets the following transcripts in Mouse: NM_009480, AK142437, BC049784.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of transcription from RNA polymerase II promoter by glucose
response to hypoxia
glucose metabolic process
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to UV
late viral transcription
cellular response to insulin stimulus
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
lipid homeostasis
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
enzyme binding
protein kinase binding
protein homodimerization activity
histone deacetylase binding
bHLH transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-qjqx6:80/100.66.79.213:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline