This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KCNQ1 SIRNA Assays ›
Gene Symbol:
Kcnq1
Gene Name:
potassium channel, voltage-gated KQT-like subfamily Q, member 1
Gene Aliases:
Kvlqt1
Chromosome Location:
Chr.1: 216293087 - 216630339 on Build Rnor_6.0
UniGene ID:
Rn.9779
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 54535

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnq1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032073.1 NP_114462.1 8 1176
GenBank mRNA AJ133685.1 - 8 1176
AY043394.1 - 7 916
U92655.1 - 7 687

Back To Top

More Information


Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of gene expression by genetic imprinting
potassium ion transport
male gonad development
positive regulation of heart rate
gene silencing
regulation of ion transmembrane transport
cellular response to drug
regulation of membrane potential
negative regulation of insulin secretion
cardiac muscle contraction
regulation of membrane repolarization
regulation of ventricular cardiac muscle cell membrane repolarization
regulation of atrial cardiac muscle cell membrane repolarization
positive regulation of cardiac muscle contraction
positive regulation of gastric acid secretion
cellular response to cAMP
potassium ion export
potassium ion transmembrane transport
response to anesthetic
cardiovascular system development
ventricular cardiac muscle cell action potential
membrane repolarization during action potential
membrane repolarization during cardiac muscle cell action potential
atrial cardiac muscle cell action potential
regulation of heart rate by cardiac conduction
potassium ion export across plasma membrane
positive regulation of potassium ion transmembrane transport
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity
protein binding
calmodulin binding
protein phosphatase 1 binding
outward rectifier potassium channel activity
protein kinase A catalytic subunit binding
protein kinase A regulatory subunit binding
protein homodimerization activity
ion channel binding
voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
voltage-gated potassium channel activity involved in atrial cardiac muscle cell action potential repolarization
scaffold protein binding
voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0