This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MTERF SIRNA Assays ›
Gene Symbol:
Mterf
Gene Name:
mitochondrial transcription termination factor
Gene Aliases:
Mterf1
Chromosome Location:
Chr.4: 27365310 - 27372374 on Build Rnor_6.0
UniGene ID:
Rn.64629
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55001

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mterf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053499.1 NP_445951.1 3 360
XM_006236069.2 XP_006236131.1 4 475
XM_006236070.2 XP_006236132.1 4 472
XM_006236071.2 XP_006236133.1 3 347
XM_006236072.2 XP_006236134.1 3 344
XM_006236073.2 XP_006236135.1 2 298
GenBank mRNA AJ292524.1 - 3 360

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA-templated transcription, termination
regulation of transcription, DNA-templated
termination of mitochondrial transcription
DNA geometric change
double-stranded DNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline