siRNA Details
| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053407.3 | NP_445859.2 | 8 | 617 | ||
| GenBank mRNA | AF214647.1 | - | 8 | 597 | ||
| BC061540.1 | - | 8 | 620 | 6919949 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asah1| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053407.3 | NP_445859.2 | 8 | 617 | ||
| GenBank mRNA | AF214647.1 | - | 8 | 597 | ||
| BC061540.1 | - | 8 | 620 | 6919949 | ||