This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ARHGEF7 SIRNA Assays ›
Gene Symbol:
Arhgef7
Gene Name:
Rho guanine nucleotide exchange factor (GEF) 7
Gene Aliases:
P85spr, Pak3bp
Chromosome Location:
Chr.16: 83006732 - 83117065 on Build Rnor_6.0
UniGene ID:
Rn.146763
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55524

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arhgef7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113521.1 NP_001106993.1 4 510
NM_001113522.1 NP_001106994.1 4 510
NM_053740.2 NP_446192.2 1 144
XM_008771376.1 XP_008769598.1 4 11482
XM_008771377.1 XP_008769599.1 4 11481
XM_008771378.1 XP_008769600.1 4 11482
XM_008771379.1 XP_008769601.1 567
XM_008771380.1 XP_008769602.1 480
GenBank mRNA AF044673.1 - 1 144
AY034823.1 - 1 144
AY996221.1 - 1 144
BC131844.1 - 2 197

Back To Top

More Information


Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
small GTPase mediated signal transduction
positive regulation of fibroblast migration
lamellipodium assembly
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
astrocyte cell migration
focal adhesion assembly
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of lamellipodium morphogenesis
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-cfg6s:80/100.66.76.62:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0