This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SEC14L2 SIRNA Assays ›
Gene Symbol:
Sec14l2
Gene Name:
SEC14-like 2 (S. cerevisiae)
Gene Aliases:
Spf
Chromosome Location:
Chr.14: 84335594 - 84360135 on Build Rnor_6.0
UniGene ID:
Rn.46850
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55738

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sec14l2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053801.2 NP_446253.2 4 254
XM_006251154.2 XP_006251216.1 4 243
GenBank mRNA AF309558.1 - 4 211
BC089785.1 - 4 254 7322689

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144520, BC005759.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transport
positive regulation of catalytic activity
positive regulation of cholesterol biosynthetic process
transporter activity
enzyme activator activity
lipid binding
transferase activity, transferring alkyl or aryl (other than methyl) groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7p2bx:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline