This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAPKAPK3 SIRNA Assays ›
Gene Symbol:
MAPKAPK3
Gene Name:
mitogen-activated protein kinase-activated protein kinase 3
Gene Aliases:
3PK, MAPKAP-K3, MAPKAP3, MAPKAPK-3, MK-3
Chromosome Location:
Chr.3: 50611862 - 50649297 on Build GRCh38
UniGene ID:
Hs.234521
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 562

Purification
Size
Availability Inventoried
Catalog # AM51331
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAPKAPK3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243925.1 NP_001230854.1 4 538
NM_001243926.1 NP_001230855.1 6 1017
NM_004635.4 NP_004626.1 4 523
GenBank mRNA AB451303.1 - 3 375
AB451434.1 - 3 375
AK290969.1 - 4 494
AK313565.1 - 3 440
BC001662.2 - 4 484 2988489
BC007591.1 - 4 460 3345889
BC010407.2 - 4 482 3833667
BC068497.1 - 6 971
U09578.1 - 4 466
U43784.1 - 3 397

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
toll-like receptor signaling pathway
response to stress
signal transduction
cell surface receptor signaling pathway
Ras protein signal transduction
peptidyl-serine phosphorylation
response to lipopolysaccharide
response to cytokine
macropinocytosis
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
MAP kinase kinase activity
protein binding
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fh6lz:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline