This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GRIFIN SIRNA Assays ›
Gene Symbol:
Grifin
Gene Name:
galectin-related inter-fiber protein
Gene Aliases:
-
Chromosome Location:
Chr.12: 16170108 - 16172144 on Build Rnor_6.0
UniGene ID:
Rn.26894
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56391

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Grifin
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057187.2 NP_476535.1 4 311
XM_008768938.1 XP_008767160.1 4 365
GenBank mRNA AF082160.1 - 4 311

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030022, BC107324, BC107323, AK021365.

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Process(es)

carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline