siRNA Details
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133421.2 | NP_596912.1 | 18 | 3728 | ||
| GenBank mRNA | AB012133.1 | - | 18 | 3728 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Marf1| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133421.2 | NP_596912.1 | 18 | 3728 | ||
| GenBank mRNA | AB012133.1 | - | 18 | 3728 | ||