siRNA Details
| Gene Symbol: Dusp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133578.1 | NP_598262.1 | 1, 2 | 561 | ||
| GenBank mRNA | AF013144.1 | - | 1, 2 | 561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp5| Gene Symbol: Dusp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133578.1 | NP_598262.1 | 1, 2 | 561 | ||
| GenBank mRNA | AF013144.1 | - | 1, 2 | 561 | ||