This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FCER2 SIRNA Assays ›
Gene Symbol:
Fcer2
Gene Name:
Fc fragment of IgE, low affinity II, receptor for (CD23)
Gene Aliases:
CD23, Fcer2a
Chromosome Location:
Chr.12: 2233772 - 2245324 on Build Rnor_6.0
UniGene ID:
Rn.10326
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 57219

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fcer2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033924.1 NP_001029096.1 9 623
NM_133550.2 NP_598234.2 8 560
XM_008768939.1 XP_008767161.1 8 787
GenBank mRNA AF381978.1 - 8 509
BC078749.1 - 9 623 7124300
X73579.1 - 7 443

Back To Top

More Information


Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s)

positive regulation of humoral immune response mediated by circulating immunoglobulin
positive regulation of nitric-oxide synthase activity
positive regulation of killing of cells of other organism
positive regulation of nitric-oxide synthase biosynthetic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-pvqqf:80/100.66.77.74:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0