This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AAGAB SIRNA Assays ›
Gene Symbol:
Aagab
Gene Name:
alpha- and gamma-adaptin binding protein
Gene Aliases:
P34
Chromosome Location:
Chr.8: 68526063 - 68564604 on Build Rnor_6.0
UniGene ID:
Rn.163023
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 57407

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aagab
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134398.2 NP_599225.1 3 366
XM_006243223.2 XP_006243285.1 3 398
GenBank mRNA AF178669.1 - 3 368
FQ224819.1 - 3 341

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025857, AK159897, AK214885, AK215257, AK209997, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein transport
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline