This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CDK12 SIRNA Assays ›
Gene Symbol:
Cdk12
Gene Name:
cyclin-dependent kinase 12
Gene Aliases:
Crk7, Crkrs, Pksc
Chromosome Location:
Chr.10: 86157188 - 86222535 on Build Rnor_6.0
UniGene ID:
Rn.48817
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 57737

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdk12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033867.1 NP_001029039.1 1 463
NM_138916.2 NP_620271.1 1 463
XM_006247250.2 XP_006247312.1 1 1017
XM_006247251.2 XP_006247313.1 1 1017
XM_006247253.2 XP_006247315.1 1 1016
GenBank mRNA AY072294.1 - 1 463
AY962568.1 - 1 463

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
regulation of MAP kinase activity
regulation of RNA splicing
protein autophosphorylation
regulation of cell cycle
phosphorylation of RNA polymerase II C-terminal domain
protein kinase activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding
RNA polymerase II carboxy-terminal domain kinase activity
cyclin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hn7z:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline