This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PZP SIRNA Assays ›
Gene Symbol:
Pzp
Gene Name:
pregnancy-zone protein
Gene Aliases:
A1m
Chromosome Location:
Chr.4: 161804792 - 161850875 on Build Rnor_6.0
UniGene ID:
Rn.11295
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58422

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pzp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145779.2 NP_665722.2 6, 7 717
XM_008763310.1 XP_008761532.1 6, 7 728
GenBank mRNA FQ209918.1 - 6, 7 746
FQ210288.1 - 6, 7 746
M77183.1 - 6, 7 713
M84000.1 - 6, 7 756

Back To Top

More Information


Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

embryo implantation
negative regulation of endopeptidase activity
serine-type endopeptidase inhibitor activity
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline