This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RWDD1 SIRNA Assays ›
Gene Symbol:
Rwdd1
Gene Name:
RWD domain containing 1
Gene Aliases:
sarip
Chromosome Location:
Chr.20: 27535316 - 27552225 on Build Rnor_6.0
UniGene ID:
Rn.234293
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58441

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rwdd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147146.2 NP_671487.1 5 668
XM_008772869.1 XP_008771091.1 5 800
GenBank mRNA FQ218614.1 - 2 203
FQ220470.1 - 2 203

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: AF092134.
This siRNA also targets the following transcripts in Mouse: NM_025614, AK005278, AK011277, AK003262, AF503942, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline