This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CDC20 SIRNA Assays ›
Gene Symbol:
Cdc20
Gene Name:
cell division cycle 20
Gene Aliases:
p55cdc
Chromosome Location:
Chr.5: 137260728 - 137264931 on Build Rnor_6.0
UniGene ID:
Rn.9262
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58709

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdc20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_171993.1 NP_741990.1 3, 4 627
GenBank mRNA BC085691.1 - 3, 4 611 7134739
U05341.1 - 3, 4 627

Back To Top

More Information


Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

mitotic nuclear division
positive regulation of cell proliferation
protein ubiquitination
anaphase-promoting complex-dependent catabolic process
positive regulation of synaptic plasticity
regulation of meiotic nuclear division
regulation of dendrite development
cell division
positive regulation of synapse maturation
positive regulation of ubiquitin protein ligase activity
protein binding
protein C-terminus binding
enzyme binding
histone deacetylase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline