This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CYP27A1 SIRNA Assays ›
Gene Symbol:
Cyp27a1
Gene Name:
cytochrome P450, family 27, subfamily a, polypeptide 1
Gene Aliases:
Cyp27, P450C27
Chromosome Location:
Chr.9: 81968289 - 81998213 on Build Rnor_6.0
UniGene ID:
Rn.94956
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58992

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyp27a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245175.1 XP_006245237.1 2 291
GenBank mRNA BC061848.1 - 2 455 5621918
M38566.1 - 2 456

Back To Top

More Information


Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

steroid catabolic process
cholesterol metabolic process
calcitriol biosynthetic process from calciol
oxidation-reduction process
iron ion binding
heme binding
vitamin D3 25-hydroxylase activity
cholesterol 26-hydroxylase activity
cholestanetriol 26-monooxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-2scln:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline