This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CCKBR SIRNA Assays ›
Gene Symbol:
Cckbr
Gene Name:
cholecystokinin B receptor
Gene Aliases:
Cck2r, Cholrec
Chromosome Location:
Chr.1: 170262218 - 170272298 on Build Rnor_6.0
UniGene ID:
Rn.90997
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 59665

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cckbr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013165.2 NP_037297.1 1, 2 201
GenBank mRNA M99418.1 - 1, 2 292
X79208.1 - 1, 2 157

Back To Top

More Information


Set Membership:

G-protein coupled receptor

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

gastric acid secretion
histamine secretion
behavioral defense response
apoptotic process
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
digestion
positive regulation of cell proliferation
positive regulation of synaptic transmission, GABAergic
response to insulin
estrous cycle
digestive tract development
gland development
positive regulation of synaptic transmission
regulation of sensory perception of pain
positive regulation of synaptic transmission, glutamatergic
ERK1 and ERK2 cascade
positive regulation of somatostatin secretion
gastrin receptor activity
type B gastrin/cholecystokinin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline