This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CCNB2 SIRNA Assays ›
Gene Symbol:
Ccnb2
Gene Name:
cyclin B2
Gene Aliases:
CycB2
Chromosome Location:
Chr.9: 70407689 - 70421554 on Build GRCm38
UniGene ID:
Mm.22592
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 60500

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccnb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007630.2 NP_031656.2 4 410
GenBank mRNA AK013312.1 - 4 379
AK076122.1 - 4 404
BC008247.1 - 4 326 3156950
CT010286.1 - 4 296
X66032.1 - 4 397

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_004701, AL080146, AK001404, AF002822, AB020981, and others.
This siRNA also targets the following transcripts in Rat: NM_001009470, XM_008758651, BC097952, BC088212, FQ228772, and others.

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s)

in utero embryonic development
cell cycle
mitotic nuclear division
growth
T cell homeostasis
thymus development
cell division

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gb242:80/100.66.79.144:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline