siRNA Details
| Gene Symbol: Dcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007831.3 | NP_031857.2 | 2 | 794 | ||
| GenBank mRNA | AK049746.1 | - | 2 | 768 | ||
| AK090377.1 | - | 2 | 795 | |||
| AK157715.1 | - | 2 | 743 | |||
| X85788.1 | - | 344 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcc| Gene Symbol: Dcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007831.3 | NP_031857.2 | 2 | 794 | ||
| GenBank mRNA | AK049746.1 | - | 2 | 768 | ||
| AK090377.1 | - | 2 | 795 | |||
| AK157715.1 | - | 2 | 743 | |||
| X85788.1 | - | 344 | ||||