This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DAF2 SIRNA Assays ›
Gene Symbol:
Daf2
Gene Name:
decay accelerating factor 2
Gene Aliases:
AI323748, Cd55b, Daf, Daf-TM, TM-DAF
Chromosome Location:
Chr.1: 130388528 - 130423037 on Build GRCm38
UniGene ID:
Mm.436759
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 61130

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Daf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007827.2 NP_031853.2 6, 7 901
GenBank mRNA BC115954.1 - 6, 7 901
L41365.1 - 6, 7 910

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune system process
complement activation, classical pathway
negative regulation of catalytic activity
innate immune response
viral entry into host cell
virus receptor activity
enzyme inhibitor activity
lipid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline