This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DAF2 SIRNA Assays ›
Gene Symbol:
Daf2
Gene Name:
decay accelerating factor 2
Gene Aliases:
AI323748, Cd55b, Daf, Daf-TM, TM-DAF
Chromosome Location:
Chr.1: 130388528 - 130423037 on Build GRCm38
UniGene ID:
Mm.436759
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 61219

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Daf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007827.2 NP_031853.2 10 1237
GenBank mRNA BC115954.1 - 10 1237
BC127150.1 - 8 908 40055987
L41365.1 - 10 1246

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune system process
complement activation, classical pathway
negative regulation of catalytic activity
innate immune response
viral entry into host cell
virus receptor activity
enzyme inhibitor activity
lipid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-24spr:80/100.66.78.83:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0