This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other F5 SIRNA Assays ›
Gene Symbol:
F5
Gene Name:
coagulation factor V
Gene Aliases:
AI173222, Cf-5, Cf5
Chromosome Location:
Chr.1: 164151859 - 164220274 on Build GRCm38
UniGene ID:
Mm.12900
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 61440

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: F5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK137521.1 - 2, 3 385
AK137537.1 - 2, 3 362
AK139240.1 - 2, 3 379
BY722729.1 - 2, 3 359
U52925.1 - 2, 3 260

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidase oxidoreductase

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-khj5q:80/100.66.76.53:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline