This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TAS2R7 SIRNA Assays ›
Gene Symbol:
TAS2R7
Gene Name:
taste 2 receptor member 7
Gene Aliases:
T2R7, TRB4
Chromosome Location:
Chr.12: 10801532 - 10802627 on Build GRCh38
UniGene ID:
Hs.533754
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 6153

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TAS2R7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023919.2 NP_076408.1 1 181
GenBank mRNA BC069091.1 - 1 124 7216905
BC093992.1 - 1 181 7939837
BC113561.1 - 1 181 8322613

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
taste receptor activity
bitter taste receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0