This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IFNGR2 SIRNA Assays ›
Gene Symbol:
Ifngr2
Gene Name:
interferon gamma receptor 2
Gene Aliases:
Ifgr2, Ifgt
Chromosome Location:
Chr.16: 91547094 - 91564007 on Build GRCm38
UniGene ID:
Mm.249364
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 62377

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ifngr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008338.3 NP_032364.1 5 701
GenBank mRNA AK137165.1 - 5 724
AK143214.1 - 5 716
AK146995.1 - 719
AK165193.1 - 5 717
AK169907.1 - 5 722
BC055745.1 - 5 701 6403454
S69336.1 - 5 725

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

cytokine-mediated signaling pathway
cytokine receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-7458x:80/100.66.72.156:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline