This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ITM2B SIRNA Assays ›
Gene Symbol:
Itm2b
Gene Name:
integral membrane protein 2B
Gene Aliases:
AI256040, Bri, Bri2, Bricd2b, D14Sel6, E25BMM, imBRI2
Chromosome Location:
Chr.14: 73362231 - 73385271 on Build GRCm38
UniGene ID:
Mm.392973
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 62438

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Itm2b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008410.2 NP_032436.1 3 522
GenBank mRNA AB030203.1 - 3 542
AB030204.1 - 2 338
AK002599.1 - 3 485
AK003224.1 - 3 489
AK003587.1 - 3 511
AK003928.1 - 3 510
AK005125.1 - 3 513
AK076139.1 - 3 531
AK077482.1 - 3 524
AK150378.1 - 3 542
AK150396.1 - 3 541
AK151714.1 - 3 541
AK151889.1 - 3 542
AK152068.1 - 3 530
AK152516.1 - 3 542
AK152650.1 - 3 542
AK152731.1 - 3 513
AK152983.1 - 3 534
AK153103.1 - 3 485
AK159307.1 - 3 485
AK159340.1 - 3 510
AK159378.1 - 3 541
AK159426.1 - 3 510
AK159619.1 - 3 534
AK159628.1 - 3 510
AK159792.1 - 3 510
AK159962.1 - 3 513
AK160101.1 - 3 510
AK160725.1 - 3 541
AK194069.1 - 87
BC004731.1 - 3 493 3583154
BC010320.1 - 3 525 3599190
BC021786.1 - 3 531 4458918
U76253.1 - 3 522

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of amyloid precursor protein biosynthetic process
extrinsic apoptotic signaling pathway in absence of ligand
beta-amyloid binding
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline