siRNA Details
| Gene Symbol: Mpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ627041.1 | - | 10 | 1001 | ||
| AK048532.1 | - | 10 | 1186 | |||
| AK081829.1 | - | 10 | 1118 | |||
| AK152379.1 | - | 10 | 1100 | |||
| AK165261.1 | - | 10 | 1119 | |||
| BC013444.1 | - | 10 | 1169 | 4221660 | ||
| BC138314.1 | - | 10 | 1059 | 8861334 | ||
| BC138315.1 | - | 10 | 1059 | 8861335 | ||
| U38196.1 | - | 10 | 1175 | |||

Mpp1
Genomic Map